Assay Details
| Gene Symbol: drp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028204.4 | NP_001023375.1 | 3 - 4 | 1608 | 81 | |
| GenBank mRNA | AF166274.1 | - | 3 - 4 | 1615 | 81 | |
More Information
Set Membership: |
|

drp-1
Genomic Map