Assay Details
| Gene Symbol: unc-77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306889.3 | NP_001293818.1 | 13 - 14 | 2308 | 115 | |
| NM_001306890.3 | NP_001293819.1 | 12 - 13 | 2128 | 115 | ||
| NM_001306891.3 | NP_001293820.1 | 13 - 14 | 2272 | 115 | ||
| NM_001306894.3 | NP_001293823.1 | 13 - 14 | 2272 | 115 | ||
| NM_001306895.3 | NP_001293824.1 | 14 - 15 | 2326 | 115 | ||
| NM_001306898.3 | NP_001293827.1 | 14 - 15 | 2326 | 115 | ||
| NM_171352.5 | NP_741413.2 | 13 - 14 | 2308 | 115 | ||
| NM_171353.6 | NP_741414.2 | 12 - 13 | 2128 | 115 | ||
More Information
Set Membership: |
|

unc-77
Genomic Map