Assay Details
| Gene Symbol: unc-77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306889.3 | NP_001293818.1 | 26 - 27 | 5284 | 79 | |
| NM_001306890.3 | NP_001293819.1 | 25 - 26 | 5104 | 79 | ||
| NM_001306892.3 | NP_001293821.1 | 25 - 26 | 5146 | 79 | ||
| NM_001306894.3 | NP_001293823.1 | 26 - 27 | 5248 | 79 | ||
| NM_001306896.3 | NP_001293825.1 | 26 - 27 | 5200 | 79 | ||
| NM_001306898.3 | NP_001293827.1 | 27 - 28 | 5302 | 79 | ||
| NM_001307702.3 | NP_001294631.1 | 25 - 26 | 5182 | 79 | ||
| NM_001307703.3 | NP_001294632.1 | 24 - 25 | 5002 | 79 | ||
More Information
Set Membership: |
|

unc-77
Genomic Map