Assay Details
| Gene Symbol: csp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380264.1 | NP_001368632.1 | 9 - 10 | 1809 | 131 | |
| NM_001380265.1 | NP_001368631.1 | 2 - 3 | 417 | 131 | ||
| NM_001383103.1 | 12 - 13 | 2111 | 131 | |||
| GenBank mRNA | AF088288.1 | - | 12 - 13 | 2108 | 131 | |
| AF088289.1 | - | 2 - 3 | 417 | 131 | ||
More Information
Set Membership: |
|

csp-2
Genomic Map