Assay Details
| Gene Symbol: deb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306814.4 | NP_001293743.1 | 13 - 14 | 2881 | 91 | |
| NM_001306815.3 | NP_001293744.1 | 13 - 14 | 2881 | 91 | ||
| NM_001306816.3 | NP_001293745.1 | 3 - 4 | 718 | 91 | ||
| NM_001380304.1 | NP_001368387.1 | 1 - 2 | 307 | 91 | ||
| NM_001380305.1 | NP_001368388.1 | 1 - 2 | 307 | 91 | ||
| NM_068703.7 | NP_501104.2 | 12 - 13 | 2641 | 91 | ||
| NM_068704.6 | NP_501105.2 | 3 - 4 | 728 | 91 | ||
| NM_171374.6 | NP_741437.1 | 12 - 13 | 2641 | 91 | ||
More Information
Set Membership: |
|

deb-1
Genomic Map