Assay Details
| Gene Symbol: deb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306815.3 | NP_001293744.1 | 15 - 16 | 3069 | 109 | |
| NM_001306816.3 | NP_001293745.1 | 5 - 6 | 906 | 109 | ||
| NM_001380304.1 | NP_001368387.1 | 3 - 4 | 495 | 109 | ||
| NM_171374.6 | NP_741437.1 | 14 - 15 | 2829 | 109 | ||
More Information
Set Membership: |
|

deb-1
Genomic Map