Assay Details
| Gene Symbol: deb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068703.7 | NP_501104.2 | 1 - 2 | 52 | 88 | |
| NM_171374.6 | NP_741437.1 | 1 - 2 | 52 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

deb-1| Gene Symbol: deb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068703.7 | NP_501104.2 | 1 - 2 | 52 | 88 | |
| NM_171374.6 | NP_741437.1 | 1 - 2 | 52 | 88 | ||
Set Membership: |
|