Assay Details
| Gene Symbol: deb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306814.4 | NP_001293743.1 | 8 - 9 | 1711 | 72 | |
| NM_001306815.3 | NP_001293744.1 | 8 - 9 | 1711 | 72 | ||
| NM_068703.7 | NP_501104.2 | 7 - 8 | 1471 | 72 | ||
| NM_171374.6 | NP_741437.1 | 7 - 8 | 1471 | 72 | ||
More Information
Set Membership: |
|

deb-1
Genomic Map