Assay Details
| Gene Symbol: pigo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059349.6 | NP_491750.3 | 6 - 7 | 723 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pigo-1| Gene Symbol: pigo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059349.6 | NP_491750.3 | 6 - 7 | 723 | 148 | |
Set Membership: |
|