Assay Details
| Gene Symbol: nmrk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059362.5 | NP_491763.1 | 4 - 5 | 491 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nmrk-1| Gene Symbol: nmrk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059362.5 | NP_491763.1 | 4 - 5 | 491 | 92 | |
Set Membership: |
|