Assay Details
| Gene Symbol: chn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059380.8 | NP_491781.2 | 4 - 5 | 276 | 105 | |
| GenBank mRNA | AF303269.1 | - | 3 - 4 | 221 | 105 | |
More Information
Set Membership: |
|

chn-1
Genomic Map