Assay Details
| Gene Symbol: T09B4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059381.6 | NP_491782.1 | 2 - 3 | 637 | 81 | |
| Gene Symbol: pigv-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059382.7 | NP_491783.1 | 2 - 3 | 630 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_059382.7 transcripts.
Set Membership: |
|

T09B4.2
Genomic Map