Assay Details
| Gene Symbol: fars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380481.1 | NP_001367677.1 | 8 - 9 | 1482 | 65 | |
| NM_059391.5 | NP_491792.1 | 8 - 9 | 1465 | 65 | ||
| Gene Symbol: ech-1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059389.5 | NP_491790.1 | 6 - 7 | 1294 | 65 | |
| Gene Symbol: mrpl-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380479.1 | NP_001367676.1 | 8 - 9 | 1466 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380479.1, NM_059389.5 transcripts.
Set Membership: |
|

fars-1
Genomic Map