Assay Details
| Gene Symbol: lpd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059405.6 | NP_491806.2 | 4 - 5 | 423 | 79 | |
| Gene Symbol: let-611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059406.7 | NP_491807.1 | 1 - 2 | 128 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_059406.7 transcripts.
Set Membership: |
|

lpd-2
Genomic Map