Assay Details
| Gene Symbol: gld-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026673.3 | NP_001021844.1 | 7 - 8 | 2307 | 62 | |
| NM_001313548.3 | NP_001300477.1 | 11 - 12 | 2176 | 62 | ||
| NM_001313549.3 | NP_001300478.1 | 9 - 10 | 1897 | 62 | ||
| NM_059440.5 | NP_491841.2 | 10 - 11 | 2780 | 62 | ||
| NM_059441.5 | NP_491842.2 | 18 - 19 | 3190 | 62 | ||
| GenBank mRNA | AY125085.1 | - | 17 - 18 | 3038 | 62 | |
More Information
Set Membership: |
|

gld-2
Genomic Map