Assay Details
| Gene Symbol: gld-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026673.3 | NP_001021844.1 | 5 - 6 | 1530 | 160 | |
| NM_001313548.3 | NP_001300477.1 | 9 - 10 | 1399 | 160 | ||
| NM_001313549.3 | NP_001300478.1 | 7 - 8 | 1120 | 160 | ||
| NM_059440.5 | NP_491841.2 | 8 - 9 | 2003 | 160 | ||
| NM_059441.5 | NP_491842.2 | 16 - 17 | 2413 | 160 | ||
| GenBank mRNA | AY125085.1 | - | 15 - 16 | 2261 | 160 | |
More Information
Set Membership: |
|

gld-2
Genomic Map