Assay Details
| Gene Symbol: gld-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059441.5 | NP_491842.2 | 3 - 4 | 452 | 94 | |
| GenBank mRNA | AY125085.1 | - | 2 - 3 | 300 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gld-2| Gene Symbol: gld-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059441.5 | NP_491842.2 | 3 - 4 | 452 | 94 | |
| GenBank mRNA | AY125085.1 | - | 2 - 3 | 300 | 94 | |
Set Membership: |
|