Assay Details
| Gene Symbol: gld-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059441.5 | NP_491842.2 | 6 - 7 | 877 | 57 | |
| GenBank mRNA | AY125085.1 | - | 5 - 6 | 725 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gld-2| Gene Symbol: gld-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059441.5 | NP_491842.2 | 6 - 7 | 877 | 57 | |
| GenBank mRNA | AY125085.1 | - | 5 - 6 | 725 | 57 | |
Set Membership: |
|