Assay Details
| Gene Symbol: gld-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059441.5 | NP_491842.2 | 7 - 8 | 1006 | 62 | |
| GenBank mRNA | AY125085.1 | - | 6 - 7 | 854 | 62 | |
More Information
Set Membership: |
|

gld-2
Genomic Map