Assay Details
| Gene Symbol: gld-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313548.3 | NP_001300477.1 | 1 - 2 | 118 | 82 | |
| NM_059441.5 | NP_491842.2 | 8 - 9 | 1132 | 82 | ||
| GenBank mRNA | AY125085.1 | - | 7 - 8 | 980 | 82 | |
More Information
Set Membership: |
|

gld-2
Genomic Map