Assay Details
| Gene Symbol: dbr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059467.6 | NP_491868.2 | 5 - 6 | 1194 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dbr-1| Gene Symbol: dbr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059467.6 | NP_491868.2 | 5 - 6 | 1194 | 129 | |
Set Membership: |
|