Assay Details
| Gene Symbol: smg-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059528.8 | NP_491929.1 | 2 - 3 | 307 | 68 | |
| GenBank mRNA | U64441.1 | - | 1 - 2 | 138 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smg-5| Gene Symbol: smg-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059528.8 | NP_491929.1 | 2 - 3 | 307 | 68 | |
| GenBank mRNA | U64441.1 | - | 1 - 2 | 138 | 68 | |
Set Membership: |
|