Assay Details
| Gene Symbol: smg-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059528.8 | NP_491929.1 | 5 - 6 | 1155 | 102 | |
| GenBank mRNA | U64441.1 | - | 4 - 5 | 986 | 102 | |
More Information
Set Membership: |
|

smg-5
Genomic Map