Assay Details
| Gene Symbol: dhod-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059529.7 | NP_491930.1 | 4 - 5 | 935 | 113 | |
| Gene Symbol: cytb-5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380600.2 | NP_001367738.1 | 3 - 4 | 849 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380600.2 transcripts.
Set Membership: |
|

dhod-1
Genomic Map