Assay Details
| Gene Symbol: dpf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059555.2 | NP_491956.1 | 14 - 15 | 2011 | 75 | |
| NM_059556.5 | NP_491957.1 | 14 - 15 | 1999 | 75 | ||
More Information
Set Membership: |
|

dpf-3
Genomic Map