Assay Details
| Gene Symbol: dpy-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059557.7 | NP_491958.1 | 2 - 3 | 203 | 137 | |
| GenBank mRNA | AY347577.1 | - | 2 - 3 | 193 | 137 | |
More Information
Set Membership: |
|

dpy-14
Genomic Map