Assay Details
| Gene Symbol: smg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025882.4 | NP_001021053.1 | 20 - 21 | 3111 | 117 | |
| NM_001374908.2 | NP_001361976.1 | 20 | 3089 | 117 | ||
| GenBank mRNA | AF149821.1 | - | 20 - 21 | 3111 | 117 | |
More Information
Set Membership: |
|

smg-1
Genomic Map