Assay Details
| Gene Symbol: bbs-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059572.6 | NP_491973.4 | 2 - 3 | 173 | 159 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bbs-9| Gene Symbol: bbs-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059572.6 | NP_491973.4 | 2 - 3 | 173 | 159 | |
Set Membership: |
|