Assay Details
| Gene Symbol: pdi-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059594.8 | NP_491995.1 | 5 - 6 | 995 | 87 | |
| GenBank mRNA | AB072567.1 | - | 5 - 6 | 997 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pdi-3| Gene Symbol: pdi-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059594.8 | NP_491995.1 | 5 - 6 | 995 | 87 | |
| GenBank mRNA | AB072567.1 | - | 5 - 6 | 997 | 87 | |
Set Membership: |
|