Assay Details
| Gene Symbol: pdi-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059594.8 | NP_491995.1 | 7 - 8 | 1300 | 80 | |
| GenBank mRNA | AB072567.1 | - | 7 - 8 | 1302 | 80 | |
More Information
Set Membership: |
|

pdi-3
Genomic Map