Assay Details
| Gene Symbol: prpf-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001263462.4 | NP_001250391.1 | 6 - 7 | 2042 | 81 | |
| NM_001263463.4 | NP_001250392.1 | 5 - 6 | 1910 | 81 | ||
More Information
Set Membership: |
|

prpf-4
Genomic Map