Assay Details
| Gene Symbol: prpf-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001263462.4 | NP_001250391.1 | 8 - 9 | 2383 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prpf-4| Gene Symbol: prpf-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001263462.4 | NP_001250391.1 | 8 - 9 | 2383 | 70 | |
Set Membership: |
|