Assay Details
| Gene Symbol: cnnm-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059639.5 | NP_492040.3 | 4 - 5 | 769 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cnnm-4| Gene Symbol: cnnm-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059639.5 | NP_492040.3 | 4 - 5 | 769 | 133 | |
Set Membership: |
|