Assay Details
| Gene Symbol: unc-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026700.2 | NP_001021871.1 | 23 - 24 | 4132 | 81 | |
| NM_001026702.3 | NP_001021873.1 | 25 - 26 | 5158 | 81 | ||
| NM_001026703.4 | NP_001021874.1 | 24 - 25 | 4141 | 81 | ||
| NM_001263573.3 | NP_001250502.1 | 24 - 25 | 4150 | 81 | ||
| NM_001306224.3 | NP_001293153.1 | 12 - 13 | 3230 | 81 | ||
| NM_001330927.3 | NP_001317864.1 | 23 - 24 | 4141 | 81 | ||
| NM_001330928.3 | NP_001317865.1 | 23 - 24 | 4096 | 81 | ||
| NM_001330929.3 | NP_001317866.1 | 23 - 24 | 4087 | 81 | ||
| NM_001330930.3 | NP_001317863.1 | 15 - 16 | 3109 | 81 | ||
| NM_001330931.3 | NP_001317867.1 | 11 - 12 | 3088 | 81 | ||
| NM_001380703.1 | NP_001367037.1 | 16 - 17 | 3118 | 81 | ||
More Information
Set Membership: |
|

unc-13
Genomic Map