Assay Details
| Gene Symbol: asic-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059698.3 | NP_492099.1 | 12 - 13 | 1536 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

asic-2| Gene Symbol: asic-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059698.3 | NP_492099.1 | 12 - 13 | 1536 | 106 | |
Set Membership: |
|