Assay Details
| Gene Symbol: F26A3.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059728.8 | NP_492129.2 | 7 - 8 | 1213 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F26A3.1| Gene Symbol: F26A3.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059728.8 | NP_492129.2 | 7 - 8 | 1213 | 86 | |
Set Membership: |
|