Assay Details
| Gene Symbol: gld-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059742.7 | NP_492143.1 | 4 - 5 | 691 | 96 | |
| Gene Symbol: T23G11.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383384.1 | 4 - 5 | 1273 | 96 | ||
More Information
Set Membership: |
|

gld-1
Genomic Map