Assay Details
| Gene Symbol: adr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373102.1 | NP_001359658.1 | 9 - 10 | 2083 | 140 | |
| NM_059752.6 | NP_492153.2 | 10 - 11 | 1987 | 140 | ||
| NM_059753.7 | NP_492154.2 | 9 - 10 | 1776 | 140 | ||
| NM_182001.4 | NP_871801.1 | 9 - 10 | 2012 | 140 | ||
| GenBank mRNA | AY150818.1 | - | 9 - 10 | 2083 | 140 | |
| AY150819.1 | - | 10 - 11 | 1816 | 140 | ||
More Information
Set Membership: |
|

adr-1
Genomic Map