Assay Details
| Gene Symbol: adr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373102.1 | NP_001359658.1 | 3 - 4 | 610 | 58 | |
| NM_059752.6 | NP_492153.2 | 4 - 5 | 499 | 58 | ||
| GenBank mRNA | AY150818.1 | - | 3 - 4 | 610 | 58 | |
More Information
Set Membership: |
|

adr-1
Genomic Map