Assay Details
| Gene Symbol: adr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373102.1 | NP_001359658.1 | 5 - 6 | 964 | 95 | |
| NM_059753.7 | NP_492154.2 | 5 - 6 | 657 | 95 | ||
| NM_182001.4 | NP_871801.1 | 5 - 6 | 893 | 95 | ||
| GenBank mRNA | AY150819.1 | - | 6 - 7 | 697 | 95 | |
More Information
Set Membership: |
|

adr-1
Genomic Map