Assay Details
| Gene Symbol: nlp-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059757.7 | NP_492158.1 | 2 - 3 | 131 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nlp-8| Gene Symbol: nlp-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059757.7 | NP_492158.1 | 2 - 3 | 131 | 69 | |
Set Membership: |
|