Assay Details
| Gene Symbol: spr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025822.3 | NP_001020993.1 | 2 - 3 | 1311 | 155 | |
| NM_001025823.6 | NP_001020994.1 | 2 - 3 | 1316 | 155 | ||
More Information
Set Membership: |
|

spr-4
Genomic Map