Assay Details
| Gene Symbol: ins-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383443.1 | 1 - 2 | 157 | 65 | ||
| GenBank mRNA | AB008096.1 | - | 1 - 2 | 157 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ins-18| Gene Symbol: ins-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383443.1 | 1 - 2 | 157 | 65 | ||
| GenBank mRNA | AB008096.1 | - | 1 - 2 | 157 | 65 | |
Set Membership: |
|