Assay Details
| Gene Symbol: ins-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383443.1 | 2 - 3 | 259 | 112 | ||
| GenBank mRNA | AB008096.1 | - | 2 - 3 | 259 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ins-18| Gene Symbol: ins-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383443.1 | 2 - 3 | 259 | 112 | ||
| GenBank mRNA | AB008096.1 | - | 2 - 3 | 259 | 112 | |
Set Membership: |
|