Assay Details
| Gene Symbol: gla-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001263777.3 | NP_001250706.1 | 9 - 10 | 1829 | 89 | |
| NM_059837.6 | NP_492238.2 | 9 - 10 | 1829 | 89 | ||
| NM_059838.3 | NP_492239.1 | 9 - 10 | 1900 | 89 | ||
More Information
Set Membership: |
|

gla-3
Genomic Map