Assay Details
| Gene Symbol: kri-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026692.5 | NP_001021863.1 | 3 - 4 | 287 | 151 | |
| NM_001026693.5 | NP_001021864.1 | 3 - 4 | 282 | 151 | ||
| GenBank mRNA | DQ372920.1 | - | 3 - 4 | 282 | 151 | |
More Information
Set Membership: |
|

kri-1
Genomic Map