Assay Details
| Gene Symbol: fitm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026694.6 | NP_001021865.1 | 5 - 6 | 568 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fitm-2| Gene Symbol: fitm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026694.6 | NP_001021865.1 | 5 - 6 | 568 | 114 | |
Set Membership: |
|