Assay Details
| Gene Symbol: kin-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383458.1 | 5 - 6 | 700 | 78 | ||
| NM_001383459.1 | 5 - 6 | 697 | 78 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

kin-10| Gene Symbol: kin-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383458.1 | 5 - 6 | 700 | 78 | ||
| NM_001383459.1 | 5 - 6 | 697 | 78 | |||
Set Membership: |
|