Assay Details
| Gene Symbol: mys-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059864.4 | NP_492265.1 | 6 - 7 | 1055 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mys-4| Gene Symbol: mys-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059864.4 | NP_492265.1 | 6 - 7 | 1055 | 120 | |
Set Membership: |
|