Assay Details
| Gene Symbol: lin-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025914.6 | NP_001021085.1 | 1 - 2 | 144 | 120 | |
| GenBank mRNA | U75912.1 | - | 1 - 2 | 154 | 120 | |
More Information
Set Membership: |
|

lin-28
Genomic Map